Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rprd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081293.1 | NP_001074762.1 | 6 - 7 | 951 | 88 | |
GenBank mRNA | AK083674.1 | - | 6 - 7 | 720 | 88 | |
AK129150.1 | - | 5 - 6 | 498 | 88 | ||
AK154692.1 | - | 5 - 6 | 625 | 88 | ||
BC067054.1 | - | 6 - 7 | 756 | 30542928 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |