Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0300019954.mm.1 |
NM_001111336.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300019954.mm.1 |
NM_001111336.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300019954.mm.1 |
NM_001270429.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300019954.mm.1 |
NM_008294.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300019954.mm.1 |
NM_008294.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300019954.mm.1 |
TC0300002551.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300019962.mm.1 |
NM_008294.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300019962.mm.1 |
NM_008294.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300019962.mm.1 |
TC0300002551.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300019953.mm.1 |
NM_001111336.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300019953.mm.1 |
NM_001111336.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300019953.mm.1 |
NM_001270429.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300019953.mm.1 |
NM_008294.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300019953.mm.1 |
NM_008294.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300019953.mm.1 |
TC0300002551.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300019955.mm.1 |
NM_001111336.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300019955.mm.1 |
NM_001111336.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300019955.mm.1 |
NM_001270429.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300019955.mm.1 |
NM_008294.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300019955.mm.1 |
NM_008294.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300019955.mm.1 |
TC0300002551.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300019951.mm.1 |
NM_001111336.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300019951.mm.1 |
NM_001111336.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300019951.mm.1 |
NM_001270429.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300019951.mm.1 |
NM_008294.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300019951.mm.1 |
NM_008294.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300019951.mm.1 |
TC0300002551.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300019961.mm.1 |
NM_001270429.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300019961.mm.1 |
NM_008294.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300019961.mm.1 |
NM_008294.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300019961.mm.1 |
TC0300002551.mm.1 |
MTA TC |