Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eqtn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001290623.1 | NP_001277552.1 | 1 - 2 | 174 | 146 | |
NM_027089.4 | NP_081365.2 | 1 - 2 | 174 | 146 | ||
GenBank mRNA | AB438105.1 | - | 1 - 2 | 83 | 146 | |
AB438106.1 | - | 1 - 2 | 83 | 146 | ||
AK006446.1 | - | 1 - 2 | 125 | 146 | ||
BC089517.1 | - | 1 - 2 | 141 | 6740450 | 146 | |
BY716419.1 | - | 1 - 2 | 141 | 146 | ||
DQ336137.1 | - | 1 - 2 | 125 | 146 | ||
EU828781.1 | - | 1 - 2 | 159 | 146 | ||
EU828782.1 | - | 1 - 2 | 159 | 146 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |