Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Katnal2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027721.2 | NP_081997.1 | 9 - 10 | 901 | 91 | |
GenBank mRNA | AK014074.1 | - | 10 - 11 | 1012 | 91 | |
AK017114.1 | - | 9 - 10 | 902 | 91 | ||
BC056963.1 | - | 9 - 10 | 848 | 91 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |