Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Katnal2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027721.2 | NP_081997.1 | 3 - 4 | 364 | 71 | |
GenBank mRNA | AK014074.1 | - | 4 - 5 | 481 | 71 | |
AK017114.1 | - | 3 - 4 | 365 | 71 | ||
BC056963.1 | - | 3 - 4 | 311 | 71 | ||
BC069977.1 | - | 3 - 4 | 355 | 30146310 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |