Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Katnal2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027721.2 | NP_081997.1 | 6 - 7 | 626 | 66 | |
GenBank mRNA | AK017114.1 | - | 6 - 7 | 627 | 66 | |
BC056963.1 | - | 6 - 7 | 573 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |