Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cachd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_198037.1 | NP_932154.1 | 9 - 10 | 1876 | 81 | |
GenBank mRNA | AK046646.1 | - | 1792 | 81 | ||
AK129397.1 | - | 9 - 10 | 1249 | 81 | ||
BC057356.1 | - | 162 | 81 | |||
BC058676.1 | - | 9 - 10 | 1876 | 6834428 | 81 | |
BC080313.1 | - | 160 | 81 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Ion Channel Mammalian Gene Collection Mus musculus Probe spans exons |