Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trim9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001286388.1 | NP_001273317.1 | 12 - 13 | 2640 | 66 | |
GenBank mRNA | AK049907.1 | - | 12 - 13 | 2354 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Mus musculus Probe spans exons |