Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnf14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164621.1 | NP_001158093.1 | 3 - 4 | 652 | 99 | |
NM_001164622.1 | NP_001158094.1 | 2 - 3 | 339 | 99 | ||
NM_020012.2 | NP_064396.2 | 2 - 3 | 339 | 99 | ||
GenBank mRNA | AF249667.1 | - | 2 - 3 | 285 | 99 | |
AK009783.1 | - | 2 - 3 | 294 | 99 | ||
AK010162.1 | - | 2 - 3 | 296 | 99 | ||
AK011316.2 | - | 3 - 4 | 340 | 99 | ||
AK038986.1 | - | 2 - 3 | 281 | 99 | ||
BC049079.1 | - | 3 - 4 | 652 | 99 | ||
BC054841.1 | - | 2 - 3 | 269 | 6489903 | 99 | |
BC094250.1 | - | 2 - 3 | 290 | 5720753 | 99 | |
CA315275.1 | - | 2 - 3 | 338 | 99 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |