Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnf14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164621.1 | NP_001158093.1 | 4 - 5 | 815 | 90 | |
NM_020012.2 | NP_064396.2 | 3 - 4 | 502 | 90 | ||
GenBank mRNA | AF249667.1 | - | 3 - 4 | 448 | 90 | |
AK010162.1 | - | 3 - 4 | 459 | 90 | ||
AK011316.2 | - | 4 - 5 | 503 | 90 | ||
BC049079.1 | - | 4 - 5 | 815 | 90 | ||
BC054841.1 | - | 3 - 4 | 432 | 6489903 | 90 | |
BC094250.1 | - | 3 - 4 | 453 | 5720753 | 90 | |
CA315275.1 | - | 3 - 4 | 501 | 90 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |