Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0100020280.mm.1 |
NM_001190984.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100020280.mm.1 |
NM_001190984.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100020280.mm.1 |
NM_001190985.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100020280.mm.1 |
NM_001190985.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100020280.mm.1 |
NM_021295.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100020280.mm.1 |
NM_021295.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100020280.mm.1 |
TC0100002499.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100020278.mm.1 |
NM_001190984.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100020278.mm.1 |
NM_001190984.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100020278.mm.1 |
NM_001190985.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100020278.mm.1 |
NM_001190985.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100020278.mm.1 |
NM_021295.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100020278.mm.1 |
NM_021295.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100020278.mm.1 |
TC0100002499.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100020279.mm.1 |
NM_001190984.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100020279.mm.1 |
NM_001190984.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100020279.mm.1 |
NM_001190985.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100020279.mm.1 |
NM_001190985.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100020279.mm.1 |
NM_021295.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100020279.mm.1 |
NM_021295.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100020279.mm.1 |
TC0100002499.mm.1 |
MTA TC |