Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptchd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083342.1 | NP_001076811.1 | 3 - 4 | 1425 | 90 | |
GenBank mRNA | AB235901.1 | - | 2 - 3 | 1179 | 90 | |
AK090326.1 | - | 3 - 4 | 1409 | 90 | ||
AK173152.1 | - | 2 - 3 | 868 | 90 | ||
BC070441.1 | - | 3 - 4 | 1357 | 30535318 | 90 | |
BC072569.1 | - | 3 - 4 | 1390 | 5719275 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |