Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptchd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083342.1 | NP_001076811.1 | 9 - 10 | 2310 | 67 | |
GenBank mRNA | AB235901.1 | - | 8 - 9 | 2064 | 67 | |
AK173152.1 | - | 8 - 9 | 1753 | 67 | ||
BC070441.1 | - | 9 - 10 | 2242 | 30535318 | 67 | |
BC072569.1 | - | 9 - 10 | 2275 | 5719275 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |