Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Phf8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113354.1 | NP_001106825.1 | 9 - 10 | 1890 | 75 | |
NM_177201.5 | NP_796175.1 | 9 - 10 | 1890 | 75 | ||
GenBank mRNA | AK036609.1 | - | 9 - 10 | 1264 | 75 | |
AK040943.1 | - | 9 - 10 | 1259 | 75 | ||
AK040969.1 | - | 9 - 10 | 1891 | 75 | ||
AK122447.1 | - | 8 - 9 | 981 | 75 | ||
AK134137.1 | - | 9 - 10 | 1227 | 75 | ||
AK147939.1 | - | 9 - 10 | 1277 | 75 | ||
BC138899.1 | - | 9 - 10 | 1159 | 8861921 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |