Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Xrcc5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009533.2 | NP_033559.2 | 7 - 8 | 923 | 85 | |
GenBank mRNA | AK010416.1 | - | 7 - 8 | 812 | 85 | |
AK081633.1 | - | 7 - 8 | 924 | 85 | ||
AK165470.1 | - | 7 - 8 | 891 | 85 | ||
AK167312.1 | - | 7 - 8 | 812 | 85 | ||
AK168913.1 | - | 7 - 8 | 812 | 85 | ||
AK169838.1 | - | 7 - 8 | 812 | 85 | ||
BC029218.1 | - | 7 - 8 | 795 | 85 | ||
BC051660.1 | - | 7 - 8 | 863 | 30008031 | 85 | |
X66323.1 | - | 7 - 8 | 824 | 85 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |