Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tex2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_198292.3 | NP_938034.2 | 7 - 8 | 2768 | 108 | |
GenBank mRNA | AK129434.1 | - | 7 - 8 | 2768 | 108 | |
AK168397.1 | - | 7 - 8 | 2778 | 108 | ||
BC057406.1 | - | 6 - 7 | 1178 | 5687258 | 108 | |
X99253.1 | - | 5 - 6 | 107 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |