Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Otud7b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025613.1 | NP_001020784.1 | 12 - 13 | 1737 | 127 | |
NM_001025614.1 | NP_001020785.1 | 11 - 12 | 1679 | 127 | ||
GenBank mRNA | AK044701.1 | - | 12 - 13 | 1738 | 127 | |
BC009124.1 | - | 6 - 7 | 646 | 127 | ||
BC037040.1 | - | 14 - 15 | 1723 | 127 | ||
BC141397.1 | - | 11 - 12 | 1569 | 9056010 | 127 | |
BC141398.1 | - | 11 - 12 | 1569 | 9056011 | 127 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |