Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tek | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001290549.1 | NP_001277478.1 | 13 - 14 | 2557 | 83 | |
NM_001290551.1 | NP_001277480.1 | 12 - 13 | 2404 | 83 | ||
NM_013690.3 | NP_038718.2 | 13 - 14 | 2557 | 83 | ||
GenBank mRNA | BC050824.1 | - | 12 - 13 | 2398 | 6333576 | 83 |
D13738.1 | - | 13 - 14 | 2559 | 84 | ||
X71426.1 | - | 14 - 15 | 2291 | 83 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protein Kinase Mammalian Gene Collection Mus musculus Inventoried Probe spans exons |