Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pde4dip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039376.2 | NP_001034465.2 | 7 - 8 | 911 | 98 | |
NM_001289701.1 | NP_001276630.1 | 7 - 8 | 911 | 98 | ||
NM_001289702.1 | NP_001276631.1 | 7 - 8 | 911 | 98 | ||
GenBank mRNA | AK141917.1 | - | 7 - 8 | 1037 | 98 | |
BC028274.1 | - | 7 - 8 | 921 | 98 | ||
BC141185.1 | - | 7 - 8 | 911 | 9055797 | 98 | |
BC145504.1 | - | 7 - 8 | 911 | 9054067 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |