Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0300007798.mm.1 |
NM_001161763.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300007798.mm.1 |
NM_001161763.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300007798.mm.1 |
NM_001161765.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300007798.mm.1 |
NM_001161765.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300007798.mm.1 |
NM_010232.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300007798.mm.1 |
TC0300000967.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300007797.mm.1 |
NM_001161765.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300007797.mm.1 |
NM_001161765.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300007797.mm.1 |
TC0300000967.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300007805.mm.1 |
NM_001161763.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300007805.mm.1 |
NM_001161763.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300007805.mm.1 |
NM_001161765.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300007805.mm.1 |
NM_001161765.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300007805.mm.1 |
NM_010232.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300007805.mm.1 |
TC0300000967.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300007803.mm.1 |
NM_001161763.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300007803.mm.1 |
NM_001161763.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300007803.mm.1 |
NM_001161765.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300007803.mm.1 |
NM_001161765.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300007803.mm.1 |
NM_010232.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300007803.mm.1 |
TC0300000967.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300007790.mm.1 |
NM_001161763.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300007790.mm.1 |
NM_001161763.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300007790.mm.1 |
TC0300000967.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300007801.mm.1 |
NM_001161763.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300007801.mm.1 |
NM_001161763.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300007801.mm.1 |
NM_001161765.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300007801.mm.1 |
NM_001161765.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300007801.mm.1 |
NM_010232.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300007801.mm.1 |
TC0300000967.mm.1 |
MTA TC |