Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Npas1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008718.2 | NP_032744.1 | 10 - 11 | 1413 | 93 | |
GenBank mRNA | AK158830.1 | - | 8 - 9 | 2070 | 93 | |
BC132113.1 | - | 9 - 10 | 1248 | 40130390 | 93 | |
BC137863.1 | - | 9 - 10 | 1248 | 8860882 | 93 | |
U77967.1 | - | 9 - 10 | 1281 | 93 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |