Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gls | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081081.2 | NP_001074550.1 | 12 - 13 | 1683 | 116 | |
NM_001113383.1 | NP_001106854.1 | 12 - 13 | 1683 | 116 | ||
GenBank mRNA | AK046297.1 | - | 7 - 8 | 605 | 116 | |
AK048072.1 | - | 10 - 11 | 861 | 116 | ||
AK173039.1 | - | 12 - 13 | 1683 | 116 | ||
BC025534.1 | - | 6 - 7 | 467 | 116 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |