Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gls | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081081.2 | NP_001074550.1 | 2 - 3 | 737 | 109 | |
NM_001113383.1 | NP_001106854.1 | 2 - 3 | 737 | 109 | ||
GenBank mRNA | AK173039.1 | - | 2 - 3 | 737 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Mus musculus Probe spans exons |