Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atp5a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007505.2 | NP_031531.1 | 8 - 9 | 1456 | 88 | |
GenBank mRNA | AK043976.1 | - | 8 - 9 | 1224 | 88 | |
AK076572.1 | - | 8 - 9 | 1224 | 88 | ||
AK098155.1 | - | 8 - 9 | 1234 | 88 | ||
AK146797.1 | - | 8 - 9 | 1227 | 88 | ||
AK150426.1 | - | 8 - 9 | 1227 | 88 | ||
AK150843.1 | - | 8 - 9 | 1224 | 88 | ||
AK151004.1 | - | 8 - 9 | 1224 | 88 | ||
AK151128.1 | - | 8 - 9 | 1233 | 88 | ||
AK151224.1 | - | 8 - 9 | 1231 | 88 | ||
AK151920.1 | - | 8 - 9 | 1228 | 88 | ||
AK152054.1 | - | 8 - 9 | 1235 | 88 | ||
AK152890.1 | - | 8 - 9 | 1227 | 88 | ||
AK153167.1 | - | 8 - 9 | 1228 | 88 | ||
AK153282.1 | - | 8 - 9 | 1224 | 88 | ||
AK159491.1 | - | 8 - 9 | 1224 | 88 | ||
AK159540.1 | - | 8 - 9 | 1224 | 88 | ||
AK159758.1 | - | 8 - 9 | 1227 | 88 | ||
AK160043.1 | - | 8 - 9 | 1234 | 88 | ||
AK164110.1 | - | 8 - 9 | 1227 | 88 | ||
AK164193.1 | - | 6 - 7 | 930 | 88 | ||
AK166709.1 | - | 8 - 9 | 1227 | 88 | ||
AK166812.1 | - | 8 - 9 | 1240 | 88 | ||
AK167159.1 | - | 8 - 9 | 1224 | 88 | ||
AK167863.1 | - | 8 - 9 | 1204 | 88 | ||
AK168198.1 | - | 8 - 9 | 1234 | 88 | ||
AK168369.1 | - | 8 - 9 | 1226 | 88 | ||
AK168617.1 | - | 8 - 9 | 1224 | 88 | ||
AK168879.1 | - | 8 - 9 | 1224 | 88 | ||
AK168890.1 | - | 8 - 9 | 1224 | 88 | ||
AK168932.1 | - | 8 - 9 | 1224 | 88 | ||
AK169080.1 | - | 8 - 9 | 1227 | 88 | ||
AK169084.1 | - | 8 - 9 | 1227 | 88 | ||
AK169105.1 | - | 8 - 9 | 1224 | 88 | ||
AK169142.1 | - | 8 - 9 | 1269 | 88 | ||
AK169300.1 | - | 8 - 9 | 1224 | 88 | ||
AK169308.1 | - | 8 - 9 | 1229 | 88 | ||
AK169414.1 | - | 8 - 9 | 1227 | 88 | ||
BC014854.1 | - | 8 - 9 | 1211 | 4500893 | 88 | |
BC020417.1 | - | 8 - 9 | 1211 | 88 | ||
BC055892.1 | - | 8 - 9 | 1661 | 88 | ||
L01062.1 | - | 8 - 9 | 1211 | 88 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Ion Channel Transporter Mammalian Gene Collection Mus musculus Probe spans exons |