Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cntln | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175275.4 | NP_780484.2 | 13 - 14 | 2114 | 155 | |
GenBank mRNA | AK052558.1 | - | 13 - 14 | 2034 | 155 | |
BB660086.1 | - | 3 - 4 | 283 | 155 | ||
BC024869.1 | - | 9 - 10 | 1143 | 155 | ||
CF734586.1 | - | 2 - 3 | 225 | 155 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |