Array Name |
Probe IDs |
Targets |
Target Type |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P23392702 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P23392700 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P23392699 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P23392701 |
- |
- |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2074291 |
BC017512.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2074291 |
AK150612.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2074291 |
AK165867.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2074291 |
AK144715.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2074291 |
NM_001163707.1 |
mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
242346 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
905135 |
AF305614.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
905135 |
AK090082.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
905135 |
AK152288.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
905135 |
AK167036.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
905135 |
BC003907.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
905135 |
NM_025433.3 |
refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
654644 |
AK035857.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
70793 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
71897 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P136878 |
- |
- |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2074291 |
BC017512.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2074291 |
AK150612.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2074291 |
AK165867.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2074291 |
AK144715.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2074291 |
NM_001163707.1 |
mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23392701 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23392691 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23392698 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23392700 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23392686 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23392695 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23392693 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23392707 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23392703 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23392692 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23392690 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23392699 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23392688 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23392705 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P136878 |
- |
- |
Whole Mouse Genome G4122A |
A_51_P228846 |
BC017512.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P228846 |
AF176526.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P557923 |
AK170743.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P557923 |
AK171483.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P557923 |
BC017512.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P557923 |
AK150612.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P557923 |
AK152888.1 |
mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P136458 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P136878 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P03945827 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P03945824 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P03945829 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P03445818 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P23392699 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P23392701 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P23392702 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P23392700 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23392699 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23392645 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23392624 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23392649 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23392701 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23392686 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23392748 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23392693 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23392650 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23392639 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23392609 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23392666 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23392618 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23392631 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23392766 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23392608 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23392672 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23392743 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23392606 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23392626 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23392644 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23392750 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23392741 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23392610 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23392665 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23392698 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23392635 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23392696 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23392589 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23392742 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23392700 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23392660 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23392752 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23392632 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23392754 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23392762 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23392642 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23392747 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23392629 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23392737 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23392652 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23392683 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23392736 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23392707 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23392596 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23392662 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23392727 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23392599 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23392682 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23392669 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23392622 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23392691 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23392751 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23392729 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23392690 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23392616 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23392758 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23392732 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23392678 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23392749 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23392590 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23392646 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23392759 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23392688 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23392627 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23392703 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23392764 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23392745 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23392603 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23392680 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23392674 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23392653 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23392761 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23392593 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23392615 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23392613 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23392617 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23392676 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23392657 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23392648 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23392637 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23392611 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23392654 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23392652 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23392680 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23392687 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23392663 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23392683 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03569668 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23392669 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23392690 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23392696 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23392691 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23392660 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23392693 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23392662 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23392684 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23392665 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23392672 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23392702 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P04055053 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23392695 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03321816 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23392666 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03445818 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23392707 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23392698 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23392676 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03321821 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23392699 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23392655 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23392674 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03945829 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23392670 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23392657 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23392678 |
- |
- |