Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxo38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_134136.3 | NP_598897.2 | 12 - 13 | 1797 | 65 | |
GenBank mRNA | AK031096.1 | - | 12 - 13 | 1747 | 65 | |
AK031347.1 | - | 12 - 13 | 1728 | 65 | ||
AK034761.1 | - | 7 - 8 | 981 | 65 | ||
AK089046.1 | - | 12 - 13 | 1769 | 65 | ||
AY267463.1 | - | 12 - 13 | 1730 | 65 | ||
BC056348.1 | - | 12 - 13 | 1749 | 5721407 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |