Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxo38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_134136.3 | NP_598897.2 | 13 - 14 | 1910 | 65 | |
GenBank mRNA | AK031096.1 | - | 13 - 14 | 1860 | 65 | |
AK031347.1 | - | 13 - 14 | 1841 | 65 | ||
AK034761.1 | - | 8 - 9 | 1094 | 65 | ||
AK089046.1 | - | 13 - 14 | 1882 | 65 | ||
AY267463.1 | - | 13 - 14 | 1843 | 65 | ||
BC056348.1 | - | 13 - 14 | 1862 | 5721407 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |