Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxo38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_134136.3 | NP_598897.2 | 16 - 17 | 2941 | 117 | |
GenBank mRNA | AK031096.1 | - | 16 - 17 | 2891 | 117 | |
AK031347.1 | - | 16 - 17 | 2872 | 117 | ||
AK089046.1 | - | 16 - 17 | 2910 | 117 | ||
AY267463.1 | - | 16 - 17 | 2871 | 117 | ||
BC056348.1 | - | 16 - 17 | 2893 | 5721407 | 117 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |