Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxo38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_134136.3 | NP_598897.2 | 20 - 21 | 3460 | 115 | |
GenBank mRNA | AK031096.1 | - | 20 - 21 | 3410 | 115 | |
AK031347.1 | - | 20 - 21 | 3391 | 115 | ||
AK089046.1 | - | 20 - 21 | 3429 | 115 | ||
AY267463.1 | - | 20 - 21 | 3390 | 115 | ||
BC056348.1 | - | 20 - 21 | 3412 | 5721407 | 115 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |