Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klhl28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025707.3 | NP_079983.1 | 2 - 3 | 1027 | 80 | |
GenBank mRNA | AK002637.1 | - | 2 - 3 | 994 | 80 | |
AK012778.2 | - | 2 - 3 | 1007 | 80 | ||
AK014466.1 | - | 2 - 3 | 1028 | 80 | ||
AK016422.1 | - | 2 - 3 | 969 | 80 | ||
BC018469.1 | - | 2 - 3 | 985 | 80 | ||
BC037017.1 | - | 2 - 3 | 1000 | 5347183 | 80 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |