Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atp8b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001001488.3 | NP_001001488.2 | 24 - 25 | 3337 | 61 | |
GenBank mRNA | AK157316.1 | - | 24 - 25 | 3605 | 61 | |
AY506548.1 | - | 24 - 25 | 3066 | 61 | ||
BC117946.2 | - | 24 - 25 | 3064 | 40098035 | 61 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |