Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atp8b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001001488.3 | NP_001001488.2 | 10 - 11 | 1267 | 144 | |
GenBank mRNA | AK036959.1 | - | 10 - 11 | 1188 | 144 | |
AK157316.1 | - | 10 - 11 | 1535 | 144 | ||
AY506548.1 | - | 10 - 11 | 996 | 144 | ||
BC117946.2 | - | 10 - 11 | 994 | 40098035 | 144 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Transporter Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |