Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Frem1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001198811.1 | NP_001185740.1 | 29 - 30 | 5928 | 140 | |
NM_177863.4 | NP_808531.2 | 28 - 29 | 5718 | 140 | ||
GenBank mRNA | AB160986.1 | - | 29 - 30 | 5928 | 140 | |
AJ616838.1 | - | 28 - 29 | 5718 | 140 | ||
BC037594.1 | - | 2 - 3 | 110 | 140 | ||
BC088732.1 | - | 3 - 4 | 252 | 140 | ||
BC138427.1 | - | 28 - 29 | 5629 | 8861447 | 140 | |
BC145274.1 | - | 28 - 29 | 5682 | 140 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |