Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ctnnbip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_023465.4 | NP_075954.1 | 2 - 3 | 222 | 77 | |
GenBank mRNA | AB021261.1 | - | 2 - 3 | 139 | 77 | |
AK009066.1 | - | 1 - 2 | 43 | 77 | ||
AK036181.1 | - | 2 - 3 | 223 | 77 | ||
AK048258.1 | - | 2 - 3 | 220 | 77 | ||
BC018436.1 | - | 2 - 3 | 184 | 77 | ||
BC038253.1 | - | 2 - 3 | 198 | 4974951 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |