Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0400023421.mm.1 |
TC0400002862.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400023421.mm.1 |
NM_001017427.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400023421.mm.1 |
NM_001017427.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400023420.mm.1 |
TC0400002862.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400023420.mm.1 |
NM_001017427.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400023420.mm.1 |
NM_001017427.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400023422.mm.1 |
TC0400002862.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400023422.mm.1 |
NM_001017427.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400023422.mm.1 |
NM_001017427.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400023423.mm.1 |
TC0400002862.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400023423.mm.1 |
NM_001017427.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400023423.mm.1 |
NM_001017427.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400023425.mm.1 |
TC0400002862.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400023425.mm.1 |
NM_001017427.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400023425.mm.1 |
NM_001017427.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400023419.mm.1 |
TC0400002862.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400023419.mm.1 |
NM_001017427.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400023419.mm.1 |
NM_001017427.1 |
Refseq |