Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrp8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080926.1 | NP_001074395.1 | 9 - 10 | 1301 | 67 | |
NR_033496.1 | 10 - 11 | 1679 | 67 | |||
GenBank mRNA | AJ312058.1 | - | 9 - 10 | 1147 | 67 | |
AK030143.1 | - | 6 - 7 | 811 | 67 | ||
AK038580.1 | - | 8 - 9 | 1338 | 67 | ||
BC127061.1 | - | 10 - 11 | 1054 | 67 | ||
D85463.1 | - | 10 - 11 | 1677 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |