Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpr89 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026229.4 | NP_080505.1 | 8 - 9 | 800 | 78 | |
GenBank mRNA | AK009734.1 | - | 7 - 8 | 679 | 78 | |
AK016787.1 | - | 8 - 9 | 800 | 78 | ||
AK034931.1 | - | 368 | 78 | |||
BC010729.1 | - | 8 - 9 | 793 | 3708143 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protein Kinase Mammalian Gene Collection Mus musculus Probe spans exons |