Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tcf7l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001142918.1 | NP_001136390.1 | 8 - 9 | 1462 | 86 | |
NM_001142919.1 | NP_001136391.1 | 8 - 9 | 1462 | 86 | ||
NM_001142920.1 | NP_001136392.1 | 9 - 10 | 1534 | 86 | ||
NM_001142921.1 | NP_001136393.1 | 8 - 9 | 1477 | 86 | ||
NM_001142922.1 | NP_001136394.1 | 8 - 9 | 1462 | 86 | ||
NM_001142923.1 | NP_001136395.1 | 8 - 9 | 1462 | 86 | ||
NM_001142924.1 | NP_001136396.1 | 5 - 6 | 764 | 86 | ||
NM_009333.3 | NP_033359.2 | 8 - 9 | 1462 | 86 | ||
GenBank mRNA | AF107298.1 | - | 8 - 9 | 1221 | 86 | |
AJ223070.1 | - | 8 - 9 | 940 | 86 | ||
AK035059.1 | - | 9 - 10 | 1362 | 86 | ||
AK038720.1 | - | 8 - 9 | 908 | 86 | ||
AK048536.1 | - | 5 - 6 | 577 | 86 | ||
AK052583.1 | - | 1 - 2 | 76 | 86 | ||
AK132465.1 | - | 9 - 10 | 1448 | 86 | ||
AY072035.1 | - | 8 - 9 | 1018 | 86 | ||
BC009807.1 | - | 8 - 9 | 967 | 86 | ||
BC011397.1 | - | 8 - 9 | 1441 | 86 | ||
BC052022.1 | - | 8 - 9 | 1041 | 5702449 | 86 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Mus musculus Probe spans exons |