Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spire1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_194355.2 | NP_919336.1 | 10 - 11 | 1321 | 86 | |
GenBank mRNA | BC046486.1 | - | 3 - 4 | 404 | 86 | |
BQ178805.1 | - | 2 - 3 | 214 | 86 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |