Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gnal | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010307.3 | NP_034437.1 | 1 - 2 | 411 | 87 | |
GenBank mRNA | AK032407.1 | - | 1 - 2 | 222 | 87 | |
AK158831.1 | - | 1 - 2 | 412 | 87 | ||
AY179169.1 | - | 1 - 2 | 145 | 87 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Detect Genomic DNA Mus musculus Probe spans exons |