Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008117.3 | NP_032143.1 | 1 - 2 | 155 | 115 | |
GenBank mRNA | AK019952.1 | - | 1 - 2 | 70 | 115 | |
AK019954.1 | - | 1 - 2 | 70 | 115 | ||
AK019956.1 | - | 1 - 2 | 67 | 115 | ||
AK019959.1 | - | 1 - 2 | 70 | 115 | ||
AK019961.1 | - | 1 - 2 | 70 | 115 | ||
AK030419.1 | - | 1 - 2 | 125 | 112 | ||
AK030715.1 | - | 1 - 2 | 70 | 115 | ||
BC061157.1 | - | 1 - 2 | 69 | 30250975 | 115 | |
X02891.1 | - | 1 - 2 | 69 | 115 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons |