Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adamtsl4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001301705.1 | NP_001288634.1 | 7 - 8 | 1552 | 66 | |
NM_144899.3 | NP_659148.2 | 7 - 8 | 1434 | 66 | ||
GenBank mRNA | AK155481.1 | - | 7 - 8 | 1484 | 66 | |
AY158701.1 | - | 7 - 8 | 1375 | 66 | ||
BC117925.1 | - | 6 - 7 | 1347 | 40097608 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protease Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |