Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nrd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_146150.2 | NP_666262.2 | 14 - 15 | 1737 | 126 | |
GenBank mRNA | AK031548.1 | - | 14 - 15 | 1737 | 126 | |
AK040377.1 | - | 13 - 14 | 1968 | 126 | ||
AK166470.1 | - | 12 - 13 | 126 | |||
BC023786.1 | - | 13 - 14 | 1115 | 126 | ||
BC026832.1 | - | 14 - 15 | 1787 | 4486176 | 126 | |
BC029977.1 | - | 5 - 6 | 373 | 126 | ||
BC036128.1 | - | 14 - 15 | 1855 | 4971791 | 126 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protease Mammalian Gene Collection Mus musculus Probe spans exons |