Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nrd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_146150.2 | NP_666262.2 | 1 - 2 | 360 | 76 | |
GenBank mRNA | AK031548.1 | - | 1 - 2 | 360 | 76 | |
AK166470.1 | - | 1 - 2 | 487 | 76 | ||
BC026832.1 | - | 1 - 2 | 410 | 4486176 | 76 | |
BC036128.1 | - | 1 - 2 | 478 | 4971791 | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protease Mammalian Gene Collection Mus musculus 1700 Probe spans exons |