Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Osbpl9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001134791.2 | NP_001128263.1 | 11 - 12 | 1037 | 157 | |
NM_133885.4 | NP_598646.4 | 14 - 15 | 1270 | 157 | ||
NM_173350.3 | NP_775485.2 | 12 - 13 | 1076 | 157 | ||
GenBank mRNA | AK035658.1 | - | 12 - 13 | 1025 | 157 | |
AK131984.1 | - | 14 - 15 | 1241 | 157 | ||
AK172010.1 | - | 12 - 13 | 1063 | 157 | ||
BC021507.1 | - | 10 - 11 | 898 | 5347121 | 157 | |
BC023759.1 | - | 14 - 15 | 1225 | 157 | ||
BC026927.1 | - | 12 - 13 | 959 | 5134780 | 157 | |
BC089163.1 | - | 11 - 12 | 978 | 5367603 | 157 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |