Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lman1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001172062.1 | NP_001165533.1 | 7 - 8 | 870 | 95 | |
NM_027400.3 | NP_081676.1 | 7 - 8 | 870 | 95 | ||
GenBank mRNA | AK011495.1 | - | 7 - 8 | 856 | 95 | |
AK044539.1 | - | 1 - 2 | 82 | 95 | ||
AK151599.1 | - | 7 - 8 | 857 | 95 | ||
AK151949.1 | - | 7 - 8 | 857 | 95 | ||
AK153554.1 | - | 7 - 8 | 857 | 95 | ||
BC057165.1 | - | 7 - 8 | 879 | 4948301 | 95 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |