Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klhl17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_198305.2 | NP_938047.1 | 4 - 5 | 805 | 68 | |
GenBank mRNA | AK154523.1 | - | 4 - 5 | 743 | 68 | |
AK170825.1 | - | 4 - 5 | 743 | 68 | ||
AK210057.1 | - | 1 - 2 | 228 | 68 | ||
AY423764.1 | - | 4 - 5 | 783 | 68 | ||
BC139309.1 | - | 4 - 5 | 746 | 8862331 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |