Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdk11b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007661.3 | NP_031687.2 | 15 - 16 | 1784 | 54 | |
GenBank mRNA | AK077668.1 | - | 15 - 16 | 1785 | 54 | |
AK147133.1 | - | 15 - 16 | 1762 | 54 | ||
BC025058.1 | - | 4 - 5 | 434 | 54 | ||
BC052920.1 | - | 15 - 16 | 1862 | 6489832 | 54 | |
L37092.1 | - | 14 - 15 | 2356 | 54 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protein Kinase Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |