Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0100005513.mm.1 |
NM_001159883.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100005513.mm.1 |
NM_001159883.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100005513.mm.1 |
NM_001159884.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100005513.mm.1 |
NM_001159884.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100005513.mm.1 |
NM_001159885.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100005513.mm.1 |
NM_001159885.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100005513.mm.1 |
NM_020266.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100005513.mm.1 |
NM_020266.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100005513.mm.1 |
NM_178055.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100005513.mm.1 |
NM_178055.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100005513.mm.1 |
TC0100000656.mm.1 |
MTA TC |